Product Details

SNP ID
rs10160391
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125838303 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTTATAACAGGCTTCTTCATTTT[C/T]AGGAGACAAACATATGTACTCAACA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PATE4 PubMed Links

Gene Details

Gene
PATE4
Gene Name
prostate and testis expressed 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144874.1 Intron NP_001138346.1

View Full Product Details