Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001167816.1 | Intron | NP_001161288.1 | |||
NM_015585.3 | Intron | NP_056400.3 | |||
XM_005260688.3 | Intron | XP_005260745.1 | |||
XM_005260689.2 | Intron | XP_005260746.1 | |||
XM_005260690.3 | Intron | XP_005260747.1 | |||
XM_005260691.3 | Intron | XP_005260748.1 | |||
XM_005260692.3 | Intron | XP_005260749.1 | |||
XM_005260693.3 | Intron | XP_005260750.1 | |||
XM_005260695.4 | Intron | XP_005260752.1 | |||
XM_005260696.4 | Intron | XP_005260753.1 | |||
XM_005260697.1 | Intron | XP_005260754.1 | |||
XM_011529211.1 | Intron | XP_011527513.1 | |||
XM_011529212.2 | Intron | XP_011527514.1 | |||
XM_011529213.1 | Intron | XP_011527515.1 | |||
XM_017027786.1 | Intron | XP_016883275.1 | |||
XM_017027787.1 | Intron | XP_016883276.1 | |||
XM_017027788.1 | Intron | XP_016883277.1 | |||
XM_017027789.1 | Intron | XP_016883278.1 | |||
XM_017027790.1 | Intron | XP_016883279.1 | |||
XM_017027791.1 | Intron | XP_016883280.1 | |||
XM_017027792.1 | Intron | XP_016883281.1 | |||
XM_017027793.1 | Intron | XP_016883282.1 | |||
XM_017027794.1 | Intron | XP_016883283.1 | |||
XM_017027795.1 | Intron | XP_016883284.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278625.1 | Intron | NP_001265554.1 | |||
NM_001278626.1 | Intron | NP_001265555.1 | |||
NM_001278627.1 | Intron | NP_001265556.1 | |||
NM_001278628.1 | Intron | NP_001265557.1 | |||
NM_016652.5 | Intron | NP_057736.4 |