Product Details

SNP ID
rs7539795
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:67143763 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTTGCATGTAAAAAATTTTTCT[A/G]TGAAGCTGTAGGTGACCTTGAGAAT
Phenotype
MIM: 607562
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf141 PubMed Links

Gene Details

Gene
C1orf141
Gene Name
chromosome 1 open reading frame 141
There are no transcripts associated with this gene.

Gene
IL23R
Gene Name
interleukin 23 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144701.2 Intron NP_653302.2
XM_005270516.2 Intron XP_005270573.1
XM_011540790.2 Intron XP_011539092.1
XM_011540791.2 Intron XP_011539093.1

View Full Product Details