Product Details

SNP ID
rs9609025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30636089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGCGCGCACCCTAATGACCTGG[A/G]GACTGAAGAGAAAAAAGGAACGAGG
Phenotype
MIM: 613441
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC35E4 PubMed Links

Gene Details

Gene
SLC35E4
Gene Name
solute carrier family 35 member E4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001479.3 1367 UTR 5 NP_001001479.1
NM_001318370.1 1367 UTR 5 NP_001305299.1
NM_001318371.1 1367 UTR 5 NP_001305300.1
XM_017028794.1 1367 UTR 5 XP_016884283.1
XM_017028795.1 1367 UTR 5 XP_016884284.1
Gene
TCN2
Gene Name
transcobalamin 2
There are no transcripts associated with this gene.

View Full Product Details