Product Details

SNP ID
rs7549579
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:44685278 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAATGACTTCAGATTTCTCATCAG[C/T]AACAGCAACTGGGGAGGGTGAATCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf228 PubMed Links

Gene Details

Gene
C1orf228
Gene Name
chromosome 1 open reading frame 228
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145636.1 Intron NP_001139108.1
XM_006710603.2 Intron XP_006710666.1
XM_006710604.2 Intron XP_006710667.1
XM_011541340.1 Intron XP_011539642.1
XM_011541341.1 Intron XP_011539643.1
XM_011541343.1 Intron XP_011539645.1
XM_011541345.1 Intron XP_011539647.1
XM_011541349.1 Intron XP_011539651.1
XM_011541354.1 Intron XP_011539656.1
XM_011541355.1 Intron XP_011539657.1
XM_011541356.2 Intron XP_011539658.1
XM_017001140.1 Intron XP_016856629.1
XM_017001141.1 Intron XP_016856630.1
XM_017001142.1 Intron XP_016856631.1
XM_017001143.1 Intron XP_016856632.1
XM_017001144.1 Intron XP_016856633.1
XM_017001145.1 Intron XP_016856634.1
XM_017001146.1 Intron XP_016856635.1
XM_017001147.1 Intron XP_016856636.1
XM_017001148.1 Intron XP_016856637.1

View Full Product Details