Product Details
- SNP ID
-
rs8011690
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:67825190 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGGCACAAGTGTAGCGCTCATAC[A/G]CACCTGGATTGGATGCTGGTGCTAT
- Phenotype
-
MIM: 602948
MIM: 612012
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RAD51B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79415557] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RAD51B
- Gene Name
- RAD51 paralog B
- Gene
- ZFYVE26
- Gene Name
- zinc finger FYVE-type containing 26
There are no transcripts associated with this gene.
View Full Product Details