Product Details

SNP ID
rs10021842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:97470368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTGTGTTTTATAGTGTGGGGG[G/T]AAAATTATTTTGCAAGGAAGAATTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STPG2 PubMed Links

Gene Details

Gene
STPG2
Gene Name
sperm tail PG-rich repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174952.2 Intron NP_777612.1
XM_011531886.2 Intron XP_011530188.1
XM_017008049.1 Intron XP_016863538.1
XM_017008050.1 Intron XP_016863539.1
XM_017008051.1 Intron XP_016863540.1
Gene
STPG2-AS1
Gene Name
STPG2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details