Product Details

SNP ID
rs9267553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31735395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGAGCGGCCGCTGCAATCAGAG[A/G]GGGGCTGGGTGACACCGAGAAGCCT
Phenotype
MIM: 606520 MIM: 602872 MIM: 604744 MIM: 603382
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf25 PubMed Links

Gene Details

Gene
C6orf25
Gene Name
chromosome 6 open reading frame 25
There are no transcripts associated with this gene.

Gene
CLIC1
Gene Name
chloride intracellular channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287593.1 Intron NP_001274522.1
NM_001287594.1 Intron NP_001274523.1
NM_001288.4 Intron NP_001279.2
Gene
DDAH2
Gene Name
dimethylarginine dimethylaminohydrolase 2
There are no transcripts associated with this gene.

Gene
MSH5
Gene Name
mutS homolog 5
There are no transcripts associated with this gene.

Gene
MSH5-SAPCD1
Gene Name
MSH5-SAPCD1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details