Product Details

SNP ID
rs7246131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57409403 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATTTTCAGTTTTTTCAGAGAC[A/G]GGGATCTCACTATATTGCCCAAACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF17 PubMed Links

Gene Details

Gene
ZNF17
Gene Name
zinc finger protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006959.2 152 Intron NP_008890.2
XM_017027205.1 152 UTR 5 XP_016882694.1
Gene
ZNF548
Gene Name
zinc finger protein 548
There are no transcripts associated with this gene.

View Full Product Details