Product Details

SNP ID
rs2855812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31504943 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAAAGATGGACTTACTCTTTGA[G/T]ACCCTGTCACCTGCCCACCCCAGTG
Phenotype
MIM: 602436
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MICB PubMed Links

Gene Details

Gene
MICB
Gene Name
MHC class I polypeptide-related sequence B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289160.1 Intron NP_001276089.1
NM_001289161.1 Intron NP_001276090.1
NM_005931.4 Intron NP_005922.2

View Full Product Details