Product Details

SNP ID
rs7518529
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:15857703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAAAAATTGTTTTTTTTCTACC[C/T]AGGTTGGAGTGTAATGGCATAATTA
Phenotype
MIM: 613484
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLJ37453 PubMed Links

Gene Details

Gene
FLJ37453
Gene Name
uncharacterized LOC729614
There are no transcripts associated with this gene.

Gene
SPEN
Gene Name
spen family transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015001.2 Intron NP_055816.2

View Full Product Details