Product Details

SNP ID
rs10076614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:146114348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATTATCAATATAAGCATTTAAT[C/T]ACAGTCACTCAAACCCTGGAGCAGT
Phenotype
MIM: 151350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LARS PubMed Links

Gene Details

Gene
LARS
Gene Name
leucyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317964.1 Intron NP_001304893.1
NM_001317965.1 Intron NP_001304894.1
NM_016460.3 Intron NP_057544.2
NM_020117.10 Intron NP_064502.9
XM_011537656.2 Intron XP_011535958.1
Gene
PLAC8L1
Gene Name
PLAC8 like 1
There are no transcripts associated with this gene.

View Full Product Details