Product Details
- SNP ID
-
hCV30001081
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:31057017 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGAGCTGGGCTCATGCTCACTTA[A/G]GAGAGGCTGAAGCTTAACCAGCGTC
- Phenotype
-
MIM: 102981
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADCYAP1R1
PubMed Links
Gene Details
- Gene
- ADCYAP1R1
- Gene Name
- ADCYAP receptor type I
There are no transcripts associated with this gene.
View Full Product Details