Product Details

SNP ID
rs9906747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63562333 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTATAAAGGAAAGTCCACAAACA[A/T]TATTAGAAAGTTGATCAGGAAATTT
Phenotype
MIM: 605973
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCAF7 PubMed Links

Gene Details

Gene
DCAF7
Gene Name
DDB1 and CUL4 associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005828.4 Intron NP_005819.3

View Full Product Details