Product Details

SNP ID
rs6689303
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:161111854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGATTCATAATCATATTTTCAT[C/G]GTATTGTGATAATTTGATATATTCA
Phenotype
MIM: 606841 MIM: 604618 MIM: 613466
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEDD PubMed Links

Gene Details

Gene
DEDD
Gene Name
death effector domain containing
There are no transcripts associated with this gene.

Gene
NIT1
Gene Name
nitrilase 1
There are no transcripts associated with this gene.

Gene
PFDN2
Gene Name
prefoldin subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012394.3 Intron NP_036526.2
XM_011509624.2 Intron XP_011507926.1

View Full Product Details