Product Details

SNP ID
rs10409107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50947422 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGTGTCATCCATTCTCTGCTCA[A/G]ATGCCACCTCTCCAAGGAGGCTCCC
Phenotype
MIM: 605643
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK5 PubMed Links

Gene Details

Gene
KLK5
Gene Name
kallikrein related peptidase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077491.1 Intron NP_001070959.1
NM_001077492.1 Intron NP_001070960.1
NM_012427.4 Intron NP_036559.1
XM_011526702.1 Intron XP_011525004.1
XM_011526703.2 Intron XP_011525005.1

View Full Product Details