Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001143943.1 | Intron | NP_001137415.1 | |||
NM_001290327.1 | Intron | NP_001277256.1 | |||
NM_032328.3 | Intron | NP_115704.1 | |||
XM_011544300.2 | Intron | XP_011542602.1 | |||
XM_011544301.1 | Intron | XP_011542603.1 | |||
XM_011544302.2 | Intron | XP_011542604.1 | |||
XM_017002538.1 | Intron | XP_016858027.1 | |||
XM_017002539.1 | Intron | XP_016858028.1 | |||
XM_017002540.1 | Intron | XP_016858029.1 | |||
XM_017002541.1 | Intron | XP_016858030.1 | |||
XM_017002542.1 | Intron | XP_016858031.1 | |||
XM_017002543.1 | Intron | XP_016858032.1 | |||
XM_017002544.1 | Intron | XP_016858033.1 | |||
XM_017002545.1 | Intron | XP_016858034.1 |