Product Details

SNP ID
rs10483213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41943521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCGCTCGGATACTAAGTCCTTC[A/G]ATAAGCATTCCCAATGTGCCCACTG
Phenotype
MIM: 610152
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPM PubMed Links

Gene Details

Gene
CENPM
Gene Name
centromere protein M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002876.2 Intron NP_001002876.1
NM_001110215.2 Intron NP_001103685.1
NM_001304370.1 Intron NP_001291299.1
NM_001304371.1 Intron NP_001291300.1
NM_001304372.1 Intron NP_001291301.1
NM_001304373.1 Intron NP_001291302.1
NM_024053.4 Intron NP_076958.1
XM_011530368.2 Intron XP_011528670.1
Gene
LINC00634
Gene Name
long intergenic non-protein coding RNA 634
There are no transcripts associated with this gene.

View Full Product Details