Product Details
- SNP ID
-
hCV30034629
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:7117351 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACCAGGAAGATGGGGTGGGTGTT[C/T]TGAAACACAAAGGTGAACCATGTGA
- Phenotype
-
MIM: 602209
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RREB1
PubMed Links
Gene Details
- Gene
- RREB1
- Gene Name
- ras responsive element binding protein 1
There are no transcripts associated with this gene.
View Full Product Details