Product Details

SNP ID
rs9978055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46099112 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCCTTGTTCTTGGGATAACACA[G/T]CTCTGGCTTGGAGGCCCCCTTGCAC
Phenotype
MIM: 120240
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COL6A2 PubMed Links

Gene Details

Gene
COL6A2
Gene Name
collagen type VI alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001849.3 311 Intron NP_001840.3
NM_058174.2 311 Intron NP_478054.2
NM_058175.2 311 Intron NP_478055.2
XM_011529451.1 311 UTR 5 XP_011527753.1

View Full Product Details