Product Details

SNP ID
rs9964754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:8621347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGTTTTTAGAAATGTTTAGTC[A/G]TGTACTTTGATAGAGGTATATGTTT
Phenotype
MIM: 616448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB12 PubMed Links

Gene Details

Gene
RAB12
Gene Name
RAB12, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025300.2 Intron NP_001020471.2
XM_006722300.3 Intron XP_006722363.1

View Full Product Details