Product Details

SNP ID
rs10489605
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:93886825 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTATAGATTTAAAATAAAATGAA[C/T]GCTTGTTTAAGGACTTATATGAAAA
Phenotype
MIM: 611199 MIM: 601176
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP2 PubMed Links

Gene Details

Gene
DNTTIP2
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
There are no transcripts associated with this gene.

Gene
GCLM
Gene Name
glutamate-cysteine ligase modifier subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308253.1 3359 UTR 3 NP_001295182.1
NM_002061.3 3359 UTR 3 NP_002052.1
XM_011541261.2 3359 UTR 3 XP_011539563.1
XM_017001056.1 3359 UTR 3 XP_016856545.1
XM_017001057.1 3359 UTR 3 XP_016856546.1

View Full Product Details