Product Details

SNP ID
rs5909433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18244219 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGTAAAAATGATGATGGTGTA[C/G]ACCATGCTGTGCCTAAGTTAGCATT
Phenotype
MIM: 300208
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCML2 PubMed Links

Gene Details

Gene
SCML2
Gene Name
sex comb on midleg-like 2 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006089.2 Intron NP_006080.1

View Full Product Details