Product Details

SNP ID
rs10276838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92571546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCATAAGCCTTCCCCACCCCAGTA[C/T]TGTAATAGTCACCCTTTCTCTAGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM133B PubMed Links

Gene Details

Gene
FAM133B
Gene Name
family with sequence similarity 133 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040057.2 Intron NP_001035146.1
NM_001288584.1 Intron NP_001275513.1
NM_152789.3 Intron NP_690002.2
XM_011516004.2 Intron XP_011514306.1
XM_011516005.2 Intron XP_011514307.1

View Full Product Details