Product Details

SNP ID
rs9604780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17638220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTACTGTTAACAAAAGCGGGTTA[C/G]TAGTTAAGACTTCCTTCATGCCCTG
Phenotype
MIM: 108746
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1E1 PubMed Links

Gene Details

Gene
ATP6V1E1
Gene Name
ATPase H+ transporting V1 subunit E1
There are no transcripts associated with this gene.

Gene
BCL2L13
Gene Name
BCL2 like 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270726.1 171 Intron NP_001257655.1
NM_001270727.1 171 Intron NP_001257656.1
NM_001270728.1 171 Intron NP_001257657.1
NM_001270729.1 171 Intron NP_001257658.1
NM_001270730.1 171 Intron NP_001257659.1
NM_001270731.1 171 Intron NP_001257660.1
NM_001270732.1 171 Intron NP_001257661.1
NM_001270733.1 171 Intron NP_001257662.1
NM_001270734.1 171 Intron NP_001257663.1
NM_001270735.1 171 Intron NP_001257664.1
NM_015367.3 171 Intron NP_056182.2
XM_011546119.1 171 UTR 5 XP_011544421.1
XM_011546120.1 171 UTR 5 XP_011544422.1
XM_011546121.2 171 UTR 5 XP_011544423.1
XM_011546123.1 171 Intron XP_011544425.1
XM_017028725.1 171 UTR 5 XP_016884214.1
XM_017028726.1 171 UTR 5 XP_016884215.1
XM_017028727.1 171 Intron XP_016884216.1
XM_017028728.1 171 UTR 5 XP_016884217.1
XM_017028729.1 171 Intron XP_016884218.1
XM_017028730.1 171 Intron XP_016884219.1
XM_017028731.1 171 UTR 5 XP_016884220.1
XM_017028732.1 171 Intron XP_016884221.1
XM_017028733.1 171 Intron XP_016884222.1
XM_017028734.1 171 UTR 5 XP_016884223.1

View Full Product Details