Product Details

SNP ID
rs10077827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139573668 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAGGAATCAAAGGACTCTGCTTT[C/G]CATTTAAAATGTCTTTCTCGGCCGG
Phenotype
MIM: 602962
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
UBE2D2 PubMed Links

Gene Details

Gene
UBE2D2
Gene Name
ubiquitin conjugating enzyme E2 D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003339.2 Intron NP_003330.1
NM_181838.1 Intron NP_862821.1
XM_017009820.1 Intron XP_016865309.1

View Full Product Details