Product Details

SNP ID
rs7539714
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:147640031 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAACTTCTTGGCTTACAGTAAGC[C/T]GCTGGACCCTAGTTTCCTTAAGCCC
Phenotype
MIM: 611471
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACP6 PubMed Links

Gene Details

Gene
ACP6
Gene Name
acid phosphatase 6, lysophosphatidic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323625.1 Intron NP_001310554.1
NM_016361.4 Intron NP_057445.4
XM_011509601.2 Intron XP_011507903.1
XM_017001422.1 Intron XP_016856911.1

View Full Product Details