Product Details
- SNP ID
-
rs10207844
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:61026205 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTTCATTGGGAGGGTTAATCTAA[C/T]TACCTCGACAACCATTACTTAAAAT
- Phenotype
-
MIM: 601789
MIM: 612787
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PEX13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115766022] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PEX13
- Gene Name
- peroxisomal biogenesis factor 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002618.3 |
|
Intron |
|
|
NP_002609.1 |
- Gene
- PUS10
- Gene Name
- pseudouridylate synthase 10
There are no transcripts associated with this gene.
View Full Product Details