Product Details

SNP ID
rs9884248
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:79178292 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAATGTATAAACTAGGAAAACGGC[A/G]AACAATATTTAATAATTGCGTGCTC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LINC01088 PubMed Links

Gene Details

Gene
LINC01088
Gene Name
long intergenic non-protein coding RNA 1088
There are no transcripts associated with this gene.

Gene
NAA11
Gene Name
N(alpha)-acetyltransferase 11, NatA catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032693.2 Intron NP_116082.1
XM_017008777.1 Intron XP_016864266.1
XM_017008778.1 Intron XP_016864267.1
XM_017008779.1 Intron XP_016864268.1

View Full Product Details