Product Details

SNP ID
rs10161436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116562227 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAACCCCAGAAAGCAGAGATAAA[A/G]GATAGTGGGAGAGTCCTGACAACAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP1LC3B2 PubMed Links

Gene Details

Gene
MAP1LC3B2
Gene Name
microtubule associated protein 1 light chain 3 beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085481.1 Intron NP_001078950.1
XM_011538652.2 Intron XP_011536954.1

View Full Product Details