Product Details

SNP ID
rs8178808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42175448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAGGAATACCTTCTGAGAGCC[A/G]GGGAGTGGGGAAAGGGGAAGGAGAC
Phenotype
MIM: 610469 MIM: 173370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP3M2 PubMed Links

Gene Details

Gene
AP3M2
Gene Name
adaptor related protein complex 3 mu 2 subunit
There are no transcripts associated with this gene.

Gene
PLAT
Gene Name
plasminogen activator, tissue type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000930.4 2353 UTR 3 NP_000921.1
NM_001319189.1 2353 UTR 3 NP_001306118.1
NM_033011.3 2353 UTR 3 NP_127509.1

View Full Product Details