Product Details

SNP ID
rs6700056
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:159835756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCTGCAGTTTGAGATCTGATG[C/G]TTCCTGAGGGCCAAGGCATTGCTGT
Phenotype
MIM: 606620
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf204 PubMed Links

Gene Details

Gene
C1orf204
Gene Name
chromosome 1 open reading frame 204
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134233.1 1219 UTR 3 NP_001127705.1
Gene
SLAMF8
Gene Name
SLAM family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020125.2 1219 UTR 3 NP_064510.1
XM_005245345.2 1219 Intron XP_005245402.1
XM_011509775.1 1219 Intron XP_011508077.1
XM_011509776.2 1219 Intron XP_011508078.1
XM_011509777.2 1219 UTR 3 XP_011508079.1
XM_011509778.2 1219 Intron XP_011508080.1
XM_011509779.2 1219 Intron XP_011508081.1
XM_011509780.2 1219 UTR 3 XP_011508082.2
XM_017001844.1 1219 UTR 3 XP_016857333.1
XM_017001845.1 1219 Intron XP_016857334.1
XM_017001846.1 1219 UTR 3 XP_016857335.1

View Full Product Details