Product Details

SNP ID
rs9627359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46268534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGGCCTGGAAGGATGCGAGGCT[C/G]CCGCTCTCCACCACAAGCAACGAAG
Phenotype
MIM: 604670
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PKDREJ PubMed Links

Gene Details

Gene
PKDREJ
Gene Name
polycystin (PKD) family receptor for egg jelly
There are no transcripts associated with this gene.

Gene
TTC38
Gene Name
tetratricopeptide repeat domain 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017931.2 130 Silent Mutation CTC,CTG L18L NP_060401.2
XM_011530259.1 130 UTR 5 XP_011528561.1
XM_011530260.2 130 Silent Mutation CTC,CTG L18L XP_011528562.1

View Full Product Details