Product Details

SNP ID
rs9494694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99546722 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGTCATGGTGCAGTTGCCTTGAG[A/G]GTCAGGCACGTTAAAGAGGAAAAAC
Phenotype
MIM: 123838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNC PubMed Links

Gene Details

Gene
CCNC
Gene Name
cyclin C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013399.1 Intron NP_001013417.1
NM_005190.3 Intron NP_005181.2
XM_006715594.1 Intron XP_006715657.1
XM_011536232.2 Intron XP_011534534.1
XM_017011436.1 Intron XP_016866925.1
Gene
TSTD3
Gene Name
thiosulfate sulfurtransferase (rhodanese)-like domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195131.1 Intron NP_001182060.1
XM_017010141.1 Intron XP_016865630.1

View Full Product Details