Product Details

SNP ID
rs10157203
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:164567658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACAGACATAATATAAGAAGAACT[A/T]GTAATGAGAAATGACTTTTCTTTTT
Phenotype
MIM: 176310
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PBX1 PubMed Links

Gene Details

Gene
PBX1
Gene Name
PBX homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204961.1 Intron NP_001191890.1
NM_001204963.1 Intron NP_001191892.1
NM_002585.3 Intron NP_002576.1
XM_005245228.3 Intron XP_005245285.1
XM_005245229.3 Intron XP_005245286.1
XM_011509590.1 Intron XP_011507892.1
XM_011509591.2 Intron XP_011507893.1
XM_011509592.1 Intron XP_011507894.1
XM_017001395.1 Intron XP_016856884.1
XM_017001396.1 Intron XP_016856885.1

View Full Product Details