Product Details

SNP ID
rs10241780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100390078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAATCATGATTTTGGGACTGAT[C/T]TGCCTCCTCAGGTGGAGGAGAAGGA
Phenotype
MIM: 605341
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PILRA PubMed Links

Gene Details

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013439.2 857 Silent Mutation ATC,ATT I215I NP_038467.2
NM_178272.1 857 Intron NP_840056.1
NM_178273.1 857 Intron NP_840057.1

View Full Product Details