Product Details

SNP ID
rs10107462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:67964790 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATGTGTTAGGCACTGCATATTCA[G/T]TAACTCACGACTTAGAACTATCTTC
Phenotype
MIM: 612139
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PREX2 PubMed Links

Gene Details

Gene
PREX2
Gene Name
phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024870.3 Intron NP_079146.2
NM_025170.5 Intron NP_079446.3
XM_011517612.2 Intron XP_011515914.1

View Full Product Details