Product Details
- SNP ID
-
rs6906510
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:152973990 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAAAAGCCAAAAAAAAAAAAACAAC[C/T]CCAGTTAAGTCCTATCTAATTCTAG
- Phenotype
-
MIM: 606013
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO5
PubMed Links
Gene Details
- Gene
- FBXO5
- Gene Name
- F-box protein 5
View Full Product Details