Product Details

SNP ID
rs6121328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32055792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGCTCCTAAACATTTCCATCACC[C/T]CTGAAAAAGGAAACTTTGTACCCAT
Phenotype
MIM: 142370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCK PubMed Links

Gene Details

Gene
HCK
Gene Name
HCK proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172129.1 Intron NP_001165600.1
NM_001172130.1 Intron NP_001165601.1
NM_001172131.1 Intron NP_001165602.1
NM_001172132.1 Intron NP_001165603.1
NM_001172133.1 Intron NP_001165604.1
NM_002110.3 Intron NP_002101.2

View Full Product Details