Product Details

SNP ID
rs9535765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51820133 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCTAATTTTCTATTTTATTTTTT[A/T]AAAAGCAGGTCACACACAGCCTACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00282 PubMed Links

Gene Details

Gene
LINC00282
Gene Name
long intergenic non-protein coding RNA 282
There are no transcripts associated with this gene.

View Full Product Details