Product Details

SNP ID
rs3842787
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122371228 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTGCTGTTCCTGCTCCTGCTCC[C/T]GCCGCTCCCCGTCCTGCTCGCGGAC
Phenotype
MIM: 176805
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PTGS1 PubMed Links

Gene Details

Gene
PTGS1
Gene Name
prostaglandin-endoperoxide synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000962.3 356 Missense Mutation CCG,CTG P17L NP_000953.2
NM_001271164.1 356 Missense Mutation CCG,CTG P17L NP_001258093.1
NM_001271165.1 356 Intron NP_001258094.1
NM_001271166.1 356 UTR 5 NP_001258095.1
NM_001271367.1 356 UTR 5 NP_001258296.1
NM_001271368.1 356 UTR 5 NP_001258297.1
NM_080591.2 356 Missense Mutation CCG,CTG P17L NP_542158.1
XM_005252105.3 356 UTR 5 XP_005252162.1
XM_011518875.2 356 UTR 5 XP_011517177.1
XM_011518876.2 356 UTR 5 XP_011517178.1
XM_017014929.1 356 UTR 5 XP_016870418.1

View Full Product Details