Product Details
- SNP ID
-
rs6132834
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:25453607 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGGCGTTCTCCACGTACAGCTC[C/T]TTCAGCAGCAGGTCGGACTTCGTGT
- Phenotype
-
MIM: 610608
MIM: 609580
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GINS1
PubMed Links
Gene Details
- Gene
- GINS1
- Gene Name
- GINS complex subunit 1
There are no transcripts associated with this gene.
- Gene
- NINL
- Gene Name
- ninein like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318226.1 |
2367 |
Silent Mutation |
AAA,AAG |
K982K |
NP_001305155.1 |
NM_025176.5 |
2367 |
Silent Mutation |
AAA,AAG |
K1331K |
NP_079452.3 |
XM_011529186.2 |
2367 |
Silent Mutation |
AAA,AAG |
K1419K |
XP_011527488.1 |
XM_011529187.2 |
2367 |
Silent Mutation |
AAA,AAG |
K1414K |
XP_011527489.1 |
XM_011529188.1 |
2367 |
Silent Mutation |
AAA,AAG |
K1403K |
XP_011527490.1 |
XM_011529189.1 |
2367 |
Silent Mutation |
AAA,AAG |
K1403K |
XP_011527491.1 |
XM_011529190.1 |
2367 |
Silent Mutation |
AAA,AAG |
K1336K |
XP_011527492.1 |
XM_011529191.1 |
2367 |
Silent Mutation |
AAA,AAG |
K1244K |
XP_011527493.1 |
XM_011529192.1 |
2367 |
Silent Mutation |
AAA,AAG |
K1031K |
XP_011527494.1 |
XM_011529193.1 |
2367 |
Silent Mutation |
AAA,AAG |
K987K |
XP_011527495.1 |
XM_011529194.2 |
2367 |
Silent Mutation |
AAA,AAG |
K969K |
XP_011527496.1 |
XM_011529195.2 |
2367 |
Silent Mutation |
AAA,AAG |
K912K |
XP_011527497.1 |
XM_011529197.2 |
2367 |
Silent Mutation |
AAA,AAG |
K778K |
XP_011527499.1 |
XM_011529198.1 |
2367 |
Silent Mutation |
AAA,AAG |
K760K |
XP_011527500.1 |
XM_017027732.1 |
2367 |
Silent Mutation |
AAA,AAG |
K1419K |
XP_016883221.1 |
XM_017027733.1 |
2367 |
Silent Mutation |
AAA,AAG |
K1239K |
XP_016883222.1 |
XM_017027734.1 |
2367 |
Silent Mutation |
AAA,AAG |
K845K |
XP_016883223.1 |
XM_017027735.1 |
2367 |
Silent Mutation |
AAA,AAG |
K778K |
XP_016883224.1 |
View Full Product Details