Product Details

SNP ID
rs10496447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113024482 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATTGTTTGGGAAGTGCCTTCTG[A/G]TACTGATTGAACAGAAATCTGTCAT
Phenotype
MIM: 605508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL36B PubMed Links

Gene Details

Gene
IL36B
Gene Name
interleukin 36, beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014438.4 Intron NP_055253.2
NM_173178.2 Intron NP_775270.1
XM_011510962.1 Intron XP_011509264.1

View Full Product Details