Product Details

SNP ID
rs9397006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169792555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCGCATTGTGTGCTGTGGAGTT[A/G]TTGGATCAGACCTGGATGCTGCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00242 PubMed Links

Gene Details

Gene
LINC00242
Gene Name
long intergenic non-protein coding RNA 242
There are no transcripts associated with this gene.

Gene
LINC00574
Gene Name
long intergenic non-protein coding RNA 574
There are no transcripts associated with this gene.

View Full Product Details