Product Details
- SNP ID
-
rs10483087
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:42506833 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGTGGAATCATAACTGATACTCA[A/G]AAACATGATCTTGAGTAATATTCAG
- Phenotype
-
MIM: 608094
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101930094
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76042482] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101930094
- Gene Name
- uncharacterized LOC101930094
There are no transcripts associated with this gene.
- Gene
- SLC37A1
- Gene Name
- solute carrier family 37 member 1
View Full Product Details