Product Details

SNP ID
rs7249904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44127607 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTTTTGTAGGGAGAATATTCATC[C/T]GTGGAAAGAAACCTTTTATTTTATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF225 PubMed Links

Gene Details

Gene
ZNF225
Gene Name
zinc finger protein 225
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321685.1 Intron NP_001308614.1
NM_013362.3 Intron NP_037494.2
XM_005259223.3 Intron XP_005259280.1
XM_011527285.2 Intron XP_011525587.1
XM_011527286.2 Intron XP_011525588.1

View Full Product Details