Product Details

SNP ID
rs9282760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7114263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTCAGGGGGATTCCATGGAACA[G/T]GTGGGAATGCTTGCTCTGAATCAAA
Phenotype
MIM: 147670
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
INSR PubMed Links

Gene Details

Gene
INSR
Gene Name
insulin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000208.3 7416 UTR 3 NP_000199.2
NM_001079817.2 7416 UTR 3 NP_001073285.1
XM_011527988.2 7416 UTR 3 XP_011526290.2
XM_011527989.2 7416 UTR 3 XP_011526291.2

View Full Product Details