Product Details

SNP ID
rs6492722
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94609862 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGTTCAATTTAACCACCAATGAA[C/T]GAAATTCCCAGTTGTACAGTCCAAG
Phenotype
MIM: 607787
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR180 PubMed Links

Gene Details

Gene
GPR180
Gene Name
G protein-coupled receptor 180
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_180989.5 Intron NP_851320.1

View Full Product Details