Product Details

SNP ID
rs5978485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:12139643 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTAAAACAGTAGCTAAAACTAAAT[A/C]ATCGATTCAGACATTTTTGGGTAGT
Phenotype
MIM: 300838
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FRMPD4 PubMed Links

Gene Details

Gene
FRMPD4
Gene Name
FERM and PDZ domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014728.3 Intron NP_055543.2
XM_005274632.3 Intron XP_005274689.2
XM_011545613.2 Intron XP_011543915.1
XM_017029983.1 Intron XP_016885472.1
XM_017029984.1 Intron XP_016885473.1
XM_017029985.1 Intron XP_016885474.1
XM_017029986.1 Intron XP_016885475.1

View Full Product Details