Product Details

SNP ID
rs9896507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28489843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACCACTGTCTAGTTTTTAGAAAC[A/C]GCACTAAAGTATCTATCTCCCAGTC
Phenotype
MIM: 604148
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC13A2 PubMed Links

Gene Details

Gene
SLC13A2
Gene Name
solute carrier family 13 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145975.1 Intron NP_001139447.1
NM_003984.3 Intron NP_003975.1
XM_006722165.2 Intron XP_006722228.1
XM_011525450.2 Intron XP_011523752.1
XM_011525451.1 Intron XP_011523753.1
XM_011525452.1 Intron XP_011523754.1
XM_011525453.2 Intron XP_011523755.1
XM_011525454.2 Intron XP_011523756.1

View Full Product Details